missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Difluoro-2-nitrotoluene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6303006

 View more versions of this product

Catalog No. AAH6303006


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,5-Difluoro-2-nitrotoluene
Colorless to Yellow
MFCD04973772
UN2810
DJEBTFSOEQWELL-UHFFFAOYSA-N
1,2-difluoro-4-methyl-5-nitrobenzene
2756252
98%
127371-50-0
C7H5F2NO2
5 g
4,5-difluoro-2-nitrotoluene, 2-nitro-4,5-difluorotoluene, 4,5-difluoro-2-methylnitrobenzene, 4,5-difluoro-1-methyl-2-nitrobenzene, benzene,1,2-difluoro-4-methyl-5-nitro, pubchem18518, acmc-1c6zu, 1-nitro-3,4-difluoro-6-methylbenzene, 1,2-difluoro-4-methyl-5-nitro-benzene, benzene, 1,2-difluoro-4-methyl-5-nitro
CC1=CC(=C(C=C1[N+](=O)[O-])F)F
173.119
173.12
Liquid

Chemical Identifiers

127371-50-0
173.119
DJEBTFSOEQWELL-UHFFFAOYSA-N
2756252
CC1=CC(=C(C=C1[N+](=O)[O-])F)F
C7H5F2NO2
MFCD04973772
4,5-difluoro-2-nitrotoluene, 2-nitro-4,5-difluorotoluene, 4,5-difluoro-2-methylnitrobenzene, 4,5-difluoro-1-methyl-2-nitrobenzene, benzene,1,2-difluoro-4-methyl-5-nitro, pubchem18518, acmc-1c6zu, 1-nitro-3,4-difluoro-6-methylbenzene, 1,2-difluoro-4-methyl-5-nitro-benzene, benzene, 1,2-difluoro-4-methyl-5-nitro
1,2-difluoro-4-methyl-5-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only