Learn More
4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%, Thermo Scientific Chemicals
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, 98%, C10H6Na2O8S2.2H2O, CAS Number-5808-22-0
Supplier: Thermo Scientific Chemicals 155191000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate | |
97.5 | |
>300.0°C | |
Authentic | |
Glass bottle | |
MFCD00150612 | |
11, 307 | |
chromotropic acid sodium salt, 4,5-dihydroxy-2,7-naphthalenedisulfonate, di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid, 4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g | |
HLVXFWDLRHCZEI-UHFFFAOYSA-L | |
4,5-dihydroxynaphthalene-2,7-disulfonic acid;sodium;dihydrate | |
124202444 | |
98% | |
Powder |
5808-22-0 | |
100.0 | |
Beige-Gray to White or Brown | |
98% | |
C10H6O8S2 | |
100 g | |
15, 2245 | |
Solubility in water: very soluble | |
OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O | |
318.27 | |
400.28 | |
0.02% max. (in water) |
Chemical Identifiers
5808-22-0 | |
318.27 | |
HLVXFWDLRHCZEI-UHFFFAOYSA-L | |
124202444 | |
OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O |
C10H6O8S2 | |
MFCD00150612 | |
chromotropic acid sodium salt, 4,5-dihydroxy-2,7-naphthalenedisulfonate, di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid, 4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g | |
4,5-dihydroxynaphthalene-2,7-disulfonic acid;sodium;dihydrate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only