Learn More
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 300600250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate | |
>300.0°C | |
Authentic | |
C10H6O8S2 | |
25 g | |
To nitrate: Passes Test, To formaldehyde: Passes Test | |
chromotropic acid sodium salt, 4,5-dihydroxy-2,7-naphthalenedisulfonate, di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid, 4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g | |
HLVXFWDLRHCZEI-UHFFFAOYSA-L | |
4,5-dihydroxynaphthalene-2,7-disulfonate | |
124202444 | |
Passes test | |
Powder |
5808-22-0 | |
Beige-Gray to White or Brown | |
Glass bottle | |
MFCD00150612 | |
11, 307 | |
15, 2245 | |
Solubility in water: very soluble | |
OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O | |
318.27 | |
400.28 | |
ACS Reagent |
Chemical Identifiers
5808-22-0 | |
318.27 | |
HLVXFWDLRHCZEI-UHFFFAOYSA-L | |
124202444 |
C10H6O8S2 | |
MFCD00150612 | |
chromotropic acid sodium salt, 4,5-dihydroxy-2,7-naphthalenedisulfonate, di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid, 4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g | |
OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only