missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzoic Acid 98.0+%, TCI America™

$95.06 - $292.65

Chemical Identifiers

CAS 4998-07-6
Molecular Formula C9H8NO6
Molecular Weight (g/mol) 226.17
MDL Number MFCD00014697
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
PubChem CID 78690
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D26285G
View Documents
TCI America
D26285G
5 g N/A
Only null left
 
D262825G
View Documents
TCI America
D262825G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4998-07-6
226.17
WWCMFGBGMJAJRX-UHFFFAOYSA-M
78690
C9H8NO6
MFCD00014697
4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC

Specifications

4998-07-6
C9H8NO6
5 g
WWCMFGBGMJAJRX-UHFFFAOYSA-M
226.17
227.17
Crystalline Powder
194°C
MFCD00014697
4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
78690
≥98.0% (GC,T)
4,5-Dimethoxy-2-nitrobenzoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'