Learn More
4,5-Dimethoxy-2-nitrobenzyl chloroformate, 97%, Thermo Scientific Chemicals
$259.12 - $259.12
Chemical Identifiers
CAS | 42855-00-5 |
---|---|
Molecular Formula | C10H10ClNO6 |
Molecular Weight (g/mol) | 275.641 |
MDL Number | MFCD00143507 |
InChI Key | RWWPKIOWBQFXEE-UHFFFAOYSA-N |
Synonym | 4,5-dimethoxy-2-nitrobenzyl carbonochloridate, nvoc-cl, 4,5-dimethoxy-2-nitrobenzyl chloroformate, 6-nitroveratryl chloroformate, nitroveratryl oxy chlorocarbamate, 6-nitroveratryloxycarbonyl chloride, carbonochloridic acid, 4,5-dimethoxy-2-nitrophenyl methyl ester, 4,5-dimethoxy-2-nitrophenyl methyl chloroformate, 4,5-dimethoxy-2-nitrophenyl methyl carbonochloridate, nvoc chloride |
PubChem CID | 3084878 |
IUPAC Name | (4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate |
SMILES | COC1=C(C=C(C(=C1)COC(=O)Cl)[N+](=O)[O-])OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1416703
|
Thermo Scientific Chemicals
L1416703 |
1 g |
Each for $259.12
|
|
|||||
Description
4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Dimethoxy-2-nitrobenzyl chloroformate is used as a reagent for protection of amines, for use in N-protection of bases in nucleotide synthesis, photochemical protecting groups in organic synthesis. It is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, acetone. Reacts with water.
Notes
Moisture and light sensitive. Store away from oxidizing agents, light, water/ moisture, heat, bases.
Chemical Identifiers
42855-00-5 | |
275.641 | |
RWWPKIOWBQFXEE-UHFFFAOYSA-N | |
3084878 | |
COC1=C(C=C(C(=C1)COC(=O)Cl)[N+](=O)[O-])OC |
C10H10ClNO6 | |
MFCD00143507 | |
4,5-dimethoxy-2-nitrobenzyl carbonochloridate, nvoc-cl, 4,5-dimethoxy-2-nitrobenzyl chloroformate, 6-nitroveratryl chloroformate, nitroveratryl oxy chlorocarbamate, 6-nitroveratryloxycarbonyl chloride, carbonochloridic acid, 4,5-dimethoxy-2-nitrophenyl methyl ester, 4,5-dimethoxy-2-nitrophenyl methyl chloroformate, 4,5-dimethoxy-2-nitrophenyl methyl carbonochloridate, nvoc chloride | |
(4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate |
Specifications
42855-00-5 | |
C10H10ClNO6 | |
1 g | |
2389168 | |
4,5-dimethoxy-2-nitrobenzyl carbonochloridate, nvoc-cl, 4,5-dimethoxy-2-nitrobenzyl chloroformate, 6-nitroveratryl chloroformate, nitroveratryl oxy chlorocarbamate, 6-nitroveratryloxycarbonyl chloride, carbonochloridic acid, 4,5-dimethoxy-2-nitrophenyl methyl ester, 4,5-dimethoxy-2-nitrophenyl methyl chloroformate, 4,5-dimethoxy-2-nitrophenyl methyl carbonochloridate, nvoc chloride | |
RWWPKIOWBQFXEE-UHFFFAOYSA-N | |
(4,5-dimethoxy-2-nitrophenyl)methyl carbonochloridate | |
3084878 | |
97% |
∽120°C (decomposition) | |
MFCD00143507 | |
UN3261 | |
Light and moisture sensitive | |
Soluble in methanol,acetone. Reacts with water. | |
COC1=C(C=C(C(=C1)COC(=O)Cl)[N+](=O)[O-])OC | |
275.641 | |
275.64 | |
4,5-Dimethoxy-2-nitrobenzyl chloroformate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only