missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(5-Nonyl)pyridine 98.0+%, TCI America™
Supplier: TCI America N04965ML
Specifications
| 4-(5-Nonyl)pyridine | |
| 267°C | |
| MFCD00010710 | |
| 5-(4-Pyridyl)nonane | |
| CCCCC(CCCC)C1=CC=NC=C1 | |
| 205.35 | |
| 205.35 | |
| Liquid |
| 2961-47-9 | |
| C14H23N | |
| 5 mL | |
| ATCGHKBXRIOBDX-UHFFFAOYSA-N | |
| 4-(nonan-5-yl)pyridine | |
| 76296 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 2961-47-9 | |
| 205.35 | |
| ATCGHKBXRIOBDX-UHFFFAOYSA-N | |
| 76296 | |
| CCCCC(CCCC)C1=CC=NC=C1 |
| C14H23N | |
| MFCD00010710 | |
| 5-(4-Pyridyl)nonane | |
| 4-(nonan-5-yl)pyridine |
Safety and Handling
TSCA : Yes