missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,6-Dichloro-2-(methylsulfonyl)pyrimidine 98.0+%, TCI America™
$48.79 - $161.54
Chemical Identifiers
CAS | 4489-34-3 |
---|---|
Molecular Formula | C5H4Cl2N2O2S |
Molecular Weight (g/mol) | 227.059 |
MDL Number | MFCD00672147 |
InChI Key | DROUVIKCNOHKBA-UHFFFAOYSA-N |
Synonym | 4,6-Dichloro-2-(methanesulfonyl)pyrimidine |
PubChem CID | 239685 |
IUPAC Name | 4,6-dichloro-2-methylsulfonylpyrimidine |
SMILES | CS(=O)(=O)C1=NC(=CC(=N1)Cl)Cl |
Chemical Identifiers
4489-34-3 | |
227.059 | |
DROUVIKCNOHKBA-UHFFFAOYSA-N | |
239685 | |
CS(=O)(=O)C1=NC(=CC(=N1)Cl)Cl |
C5H4Cl2N2O2S | |
MFCD00672147 | |
4,6-Dichloro-2-(methanesulfonyl)pyrimidine | |
4,6-dichloro-2-methylsulfonylpyrimidine |
Specifications
4489-34-3 | |
C5H4Cl2N2O2S | |
1 g | |
DROUVIKCNOHKBA-UHFFFAOYSA-N | |
4,6-dichloro-2-methylsulfonylpyrimidine | |
239685 | |
≥98.0% (GC) | |
4,6-Dichloro-2-(methylsulfonyl)pyrimidine |
120°C | |
MFCD00672147 | |
4,6-Dichloro-2-(methanesulfonyl)pyrimidine | |
CS(=O)(=O)C1=NC(=CC(=N1)Cl)Cl | |
227.059 | |
227.06 | |
Crystalline Powder |
Safety and Handling
TSCA : No