missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloronicotinic acid, 97%

CAS: 73027-79-9 | C6H3Cl2NO2 | 191.995 g/mol

$238.32 - $845.58

Chemical Identifiers

CAS 73027-79-9
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00234146
InChI Key ILMIEWNDXAKVNI-UHFFFAOYSA-N
Synonym 4,6-dichloronicotinic acid, 2,4-dichloro-5-carboxylpyridine, 4,6-dichloro nicotinic acid, 2,4-dichloropyridine-5-carboxylic acid, 3-pyridinecarboxylic acid, 4,6-dichloro, 2,4-dichloro-5-carboxypyridine, pubchem18521, acmc-209op3, 4.6-dichloronicotinic acid, 4,6-dichloro-nicotinic acid
PubChem CID 817158
IUPAC Name 4,6-dichloropyridine-3-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)C(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6138106
View Documents
Thermo Scientific Chemicals
H6138106
5 g
Each for $238.32
Only null left
 
AAH6138114
View Documents
Thermo Scientific Chemicals
H6138114
25 g
Each for $845.58
Only null left
 
Description

Description

4,6-Dichloronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Protect from heat. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

73027-79-9
191.995
ILMIEWNDXAKVNI-UHFFFAOYSA-N
817158
C1=C(C(=CN=C1Cl)C(=O)O)Cl
C6H3Cl2NO2
MFCD00234146
4,6-dichloronicotinic acid, 2,4-dichloro-5-carboxylpyridine, 4,6-dichloro nicotinic acid, 2,4-dichloropyridine-5-carboxylic acid, 3-pyridinecarboxylic acid, 4,6-dichloro, 2,4-dichloro-5-carboxypyridine, pubchem18521, acmc-209op3, 4.6-dichloronicotinic acid, 4,6-dichloro-nicotinic acid
4,6-dichloropyridine-3-carboxylic acid

Specifications

73027-79-9
MFCD00234146
4,6-dichloronicotinic acid, 2,4-dichloro-5-carboxylpyridine, 4,6-dichloro nicotinic acid, 2,4-dichloropyridine-5-carboxylic acid, 3-pyridinecarboxylic acid, 4,6-dichloro, 2,4-dichloro-5-carboxypyridine, pubchem18521, acmc-209op3, 4.6-dichloronicotinic acid, 4,6-dichloro-nicotinic acid
ILMIEWNDXAKVNI-UHFFFAOYSA-N
4,6-dichloropyridine-3-carboxylic acid
817158
97%
4,6-Dichloronicotinic acid
C6H3Cl2NO2
5 g
Slightly soluble in water.
C1=C(C(=CN=C1Cl)C(=O)O)Cl
191.995
192
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only