Learn More
4,7-Dibromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5630614
Description
4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9'-heptadecanyl-2,7-carbazole-alt-5,5-(4',7'-di-2-thienyl-2',1',3'-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene-2,6-diyl]] (PCPDTBT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9′-heptadecanyl-2,7-carbazole-alt-5,5-(4′,7′-di-2-thienyl-2′,1′,3′-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene-2,6-diyl]] (PCPDTBT).
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents.
Specifications
4,7-Dibromo-2,1,3-benzothiadiazole | |
186°C to 190°C | |
MFCD00658844 | |
UN2811 | |
Soluble in toluene. | |
BrC1=CC=C(Br)C2=NSN=C12 | |
293.96 | |
293.97 |
15155-41-6 | |
C6H2Br2N2S | |
25 g | |
4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole | |
FEOWHLLJXAECMU-UHFFFAOYSA-N | |
4,7-dibromo-2,1,3-benzothiadiazole | |
626361 | |
97% |
Chemical Identifiers
15155-41-6 | |
293.96 | |
FEOWHLLJXAECMU-UHFFFAOYSA-N | |
626361 | |
BrC1=CC=C(Br)C2=NSN=C12 |
C6H2Br2N2S | |
MFCD00658844 | |
4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole | |
4,7-dibromo-2,1,3-benzothiadiazole |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only