missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4',7-Dihydroxyisoflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2287706

 View more versions of this product

Catalog No. AAB2287706


Explore available promotions

Description

Description

4',7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Solubility
Insoluble in water. Soluble in DMSO (25 mg/ml), ethanol (2 mg/ml), DMF (10 mg/ml), and 1:10 DMSO:PBS (pH 7.2) (0.15 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

4',7-Dihydroxyisoflavone
328°C to 332°C (decomposition)
MFCD00016954
231523
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
5281708
254.24
486-66-8
C15H10O4
5 g
14,2801
Insoluble in water. Soluble in DMSO (25mg/ml),ethanol (2mg/ml),DMF (10mg/ml),and 1:10 DMSO:PBS (pH 7.2) (0.15mg/ml).
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
254.241
CHEBI:28197
97%

Chemical Identifiers

486-66-8
254.241
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
5281708
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O4
MFCD00016954
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
CHEBI:28197
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 207-635-4

RTECSNumber : DJ3100040

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only