Learn More
4,7-Dimethyl-1,10-phenanthroline, 98%
CAS: 3248-05-3 | C14H12N2 | 208.26 g/mol
$90.19 - $707.89
Chemical Identifiers
| CAS | 3248-05-3 |
|---|---|
| Molecular Formula | C14H12N2 |
| Molecular Weight (g/mol) | 208.26 |
| MDL Number | MFCD00004979 |
| InChI Key | JIVLDFFWTQYGSR-UHFFFAOYSA-N |
| Synonym | 4,7-dimethyl-o-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl, 4,7-dimethyl-1,10 phenanthroline, 4,7-dimethylpyridino 3,2-h quinoline, acmc-1cjn6, bis 5-chlorophenyl acetic acid, 4,7-dimethyl-1,9-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl-, hydrate |
| PubChem CID | 72792 |
| IUPAC Name | 4,7-dimethyl-1,10-phenanthroline |
| SMILES | CC1=C2C=CC3=C(C=CN=C3C2=NC=C1)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1777901
|
Thermo Scientific Chemicals
A1777901 |
0.25 g |
N/A
|
|
|||||
|
AAA1777903
|
Thermo Scientific Chemicals
A1777903 |
1 g |
N/A
|
|
|||||
|
AAA1777906
|
Thermo Scientific Chemicals
A1777906 |
5 g |
N/A
|
|
|||||
Description
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.
Solubility
Soluble in benzene and alcohol. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
| 3248-05-3 | |
| 208.26 | |
| JIVLDFFWTQYGSR-UHFFFAOYSA-N | |
| 72792 | |
| CC1=C2C=CC3=C(C=CN=C3C2=NC=C1)C |
| C14H12N2 | |
| MFCD00004979 | |
| 4,7-dimethyl-o-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl, 4,7-dimethyl-1,10 phenanthroline, 4,7-dimethylpyridino 3,2-h quinoline, acmc-1cjn6, bis 5-chlorophenyl acetic acid, 4,7-dimethyl-1,9-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl-, hydrate | |
| 4,7-dimethyl-1,10-phenanthroline |
Specifications
| 3248-05-3 | |
| C14H12N2 | |
| 0.25 g | |
| Soluble in benzene and alcohol. Slightly soluble in water. | |
| CC1=C2C=CC3=C(C=CN=C3C2=NC=C1)C | |
| 208.26 | |
| 208.26 | |
| 4,7-Dimethyl-1,10-phenanthroline |
| 193°C to 195°C | |
| MFCD00004979 | |
| 4,7-dimethyl-o-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl, 4,7-dimethyl-1,10 phenanthroline, 4,7-dimethylpyridino 3,2-h quinoline, acmc-1cjn6, bis 5-chlorophenyl acetic acid, 4,7-dimethyl-1,9-phenanthroline, 1,10-phenanthroline, 4,7-dimethyl-, hydrate | |
| JIVLDFFWTQYGSR-UHFFFAOYSA-N | |
| 4,7-dimethyl-1,10-phenanthroline | |
| 72792 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 221-827-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only