missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one 98.0+%, TCI America™

Supplier:  TCI America T24711G

 View more versions of this product

Catalog No. T24711G



Description

Description

Specifications

Specifications

4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one
181°C
C8H8O4
1 g
UAFDSOCDQRHNCL-UHFFFAOYNA-N
4,9,11-trioxatetracyclo[5.3.1.0²,⁶.0⁸,¹⁰]undecan-3-one
20632532
≥98.0% (GC)
632339-23-2
White-Yellow
MFCD10566937
2,6-Epoxy-2,2a,5,5a,6,6a-hexahydrooxireno[f]isobenzofuran-3(1aH)-one
O=C1OCC2C3OC(C4OC34)C12
168.15
168.15
Crystalline Powder

Chemical Identifiers

632339-23-2
168.15
UAFDSOCDQRHNCL-UHFFFAOYNA-N
20632532
O=C1OCC2C3OC(C4OC34)C12
C8H8O4
MFCD10566937
2,6-Epoxy-2,2a,5,5a,6,6a-hexahydrooxireno[f]isobenzofuran-3(1aH)-one
4,9,11-trioxatetracyclo[5.3.1.0²,⁶.0⁸,¹⁰]undecan-3-one
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions