missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamido-2-methylbenzoic acid, 96%, Thermo Scientific Chemicals

$108.29 - $365.23

Chemical Identifiers

CAS 103204-69-9
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD02258874
InChI Key AQPDTYYKDYMCTH-UHFFFAOYSA-N
Synonym 4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid
PubChem CID 2735224
IUPAC Name 4-acetamido-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)NC(=O)C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2717406
View Documents
Thermo Scientific Chemicals
H2717406
5 g
Each for $108.29
 
AAH2717414
View Documents
Thermo Scientific Chemicals
H2717414
25 g
Each for $365.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103204-69-9
193.202
AQPDTYYKDYMCTH-UHFFFAOYSA-N
2735224
CC1=C(C=CC(=C1)NC(=O)C)C(=O)O
C10H11NO3
MFCD02258874
4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid
4-acetamido-2-methylbenzoic acid

Specifications

103204-69-9
C10H11NO3
5 g
AQPDTYYKDYMCTH-UHFFFAOYSA-N
4-acetamido-2-methylbenzoic acid
2735224
96%
∽248°C (decomposition)
MFCD02258874
4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid
CC1=C(C=CC(=C1)NC(=O)C)C(=O)O
193.202
193.21
4-Acetamido-2-methylbenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only