missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Acetamido-2-methylbenzoic Acid 96.0+%, TCI America™
Supplier: TCI America A283825G
Specifications
4-Acetamido-2-methylbenzoic Acid | |
237°C | |
C10H11NO3 | |
25 g | |
AQPDTYYKDYMCTH-UHFFFAOYSA-N | |
4-acetamido-2-methylbenzoic acid | |
2735224 | |
≥96.0% (HPLC,T) |
103204-69-9 | |
Yellow | |
MFCD02258874 | |
4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid | |
CC1=C(C=CC(=C1)NC(=O)C)C(=O)O | |
193.202 | |
193.20 | |
Crystalline Powder |
Chemical Identifiers
103204-69-9 | |
193.202 | |
AQPDTYYKDYMCTH-UHFFFAOYSA-N | |
2735224 | |
CC1=C(C=CC(=C1)NC(=O)C)C(=O)O |
C10H11NO3 | |
MFCD02258874 | |
4-acetylamino-2-methylbenzoic acid, benzoic acid, 4-acetylamino-2-methyl, 4-acetamido-2-methyl-benzoic acid, acmc-20amqf, 4-acetamido-o-toluic acid, 4'-acetamido-2-methylbenzoicacid, 4-acetylamino-2-methyl-benzoic acid, o-toluic acid, 4-acetamido-6ci, n-acetyl-4-amino-2-methyl-benzoic acid | |
4-acetamido-2-methylbenzoic acid |
Safety and Handling
TSCA : No