missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidobenzenesulfonyl Azide 98.0+%, TCI America™

$63.21 - $600.40

Chemical Identifiers

CAS 2158-14-7
Molecular Formula C8H8N4O3S
Molecular Weight (g/mol) 240.24
MDL Number MFCD00029626
InChI Key NTMHWRHEGDRTPD-UHFFFAOYSA-N
Synonym 4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
PubChem CID 5129185
IUPAC Name 4-acetamidobenzene-1-sulfonyl azide
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A17865G
View Documents
TCI America
A17865G
5 g N/A
 
A178625G
View Documents
TCI America
A178625G
25 g N/A
 
A1786100G
View Documents
TCI America
A1786100G
100 g N/A
 
Description

Description

Specifications

Chemical Identifiers

2158-14-7
240.24
NTMHWRHEGDRTPD-UHFFFAOYSA-N
5129185
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
C8H8N4O3S
MFCD00029626
4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
4-acetamidobenzene-1-sulfonyl azide

Specifications

2158-14-7
Purple-White
MFCD00029626
4-acetamidobenzenesulfonyl azide, p-absa, n-4-azidosulfonylphenyl acetamide, 4-acetamidobenzenesulfonylazide, 4-acetamidobenzenesulphonyl azide, 4-acetamidobenzene sulfonyl azide, 4-acetamidobenzene-1-sulfonyl azide, acmc-1chyo, ksc491c8r, p-acetamidobenzensulfonyl azide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
240.24
240.24
Crystalline Powder
112°C
C8H8N4O3S
5 g
NTMHWRHEGDRTPD-UHFFFAOYSA-N
4-acetamidobenzene-1-sulfonyl azide
5129185
≥98.0% (HPLC)
4-Acetamidobenzenesulfonyl Azide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'