missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamidopiperidine 98.0+%, TCI America™
$195.73 - $583.44
Chemical Identifiers
| CAS | 5810-56-0 |
|---|---|
| Molecular Formula | C7H14N2O |
| Molecular Weight (g/mol) | 142.202 |
| MDL Number | MFCD05254805 |
| InChI Key | YLWUSMHZABTZGP-UHFFFAOYSA-N |
| Synonym | 4-acetamidopiperidine, n-piperidin-4-yl acetamide, 4-acetylamino-piperidine, 4-acetylaminopiperidine, n-piperidin-4-yl-acetamide, acetamide, n-4-piperidinyl, 4-acetylamino piperidine, n-4-piperidinyl acetamide, acmc-20aiud, pubchem9227 |
| PubChem CID | 1445156 |
| IUPAC Name | N-piperidin-4-ylacetamide |
| SMILES | CC(=O)NC1CCNCC1 |
Chemical Identifiers
| 5810-56-0 | |
| 142.202 | |
| YLWUSMHZABTZGP-UHFFFAOYSA-N | |
| 1445156 | |
| CC(=O)NC1CCNCC1 |
| C7H14N2O | |
| MFCD05254805 | |
| 4-acetamidopiperidine, n-piperidin-4-yl acetamide, 4-acetylamino-piperidine, 4-acetylaminopiperidine, n-piperidin-4-yl-acetamide, acetamide, n-4-piperidinyl, 4-acetylamino piperidine, n-4-piperidinyl acetamide, acmc-20aiud, pubchem9227 | |
| N-piperidin-4-ylacetamide |
Specifications
| 5810-56-0 | |
| White-Yellow | |
| MFCD05254805 | |
| 4-acetamidopiperidine, n-piperidin-4-yl acetamide, 4-acetylamino-piperidine, 4-acetylaminopiperidine, n-piperidin-4-yl-acetamide, acetamide, n-4-piperidinyl, 4-acetylamino piperidine, n-4-piperidinyl acetamide, acmc-20aiud, pubchem9227 | |
| CC(=O)NC1CCNCC1 | |
| 142.202 | |
| 142.20 | |
| Crystalline Powder |
| 141°C | |
| C7H14N2O | |
| 5 g | |
| YLWUSMHZABTZGP-UHFFFAOYSA-N | |
| N-piperidin-4-ylacetamide | |
| 1445156 | |
| ≥98.0% (GC,T) | |
| 4-Acetamidopiperidine |
Safety and Handling
TSCA : No