missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetoxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

$85.03 - $497.70

Chemical Identifiers

CAS 881-68-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00003362
InChI Key PZSJOBKRSVRODF-UHFFFAOYSA-N
Synonym vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
PubChem CID 61229
ChEBI CHEBI:86956
IUPAC Name (4-formyl-2-methoxyphenyl) acetate
SMILES CC(=O)OC1=C(C=C(C=C1)C=O)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1210314
View Documents
Thermo Scientific Chemicals
A1210314
25 g
Each for $85.03
 
AAA1210322
View Documents
Thermo Scientific Chemicals
A1210322
100 g
Each for $256.05
 
AAA1210330
View Documents
Thermo Scientific Chemicals
A1210330
250 g
Each for $497.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

881-68-5
194.186
PZSJOBKRSVRODF-UHFFFAOYSA-N
61229
(4-formyl-2-methoxyphenyl) acetate
C10H10O4
MFCD00003362
vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
CHEBI:86956
CC(=O)OC1=C(C=C(C=C1)C=O)OC

Specifications

881-68-5
C10H10O4
25 g
Air Sensitive
PZSJOBKRSVRODF-UHFFFAOYSA-N
(4-formyl-2-methoxyphenyl) acetate
61229
194.18
4-Acetoxy-3-methoxybenzaldehyde
74°C to 78°C
MFCD00003362
1963795
vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
CC(=O)OC1=C(C=C(C=C1)C=O)OC
194.186
CHEBI:86956
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-920-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only