missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5966003
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester | |
72°C to 74°C | |
MFCD22419259 | |
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F | |
282.094 | |
282.09 |
1022154-80-8 | |
C14H17BF2O3 | |
1 g | |
NDJMQXWRZUTNSA-UHFFFAOYSA-N | |
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone | |
68668352 | |
96% |
Chemical Identifiers
1022154-80-8 | |
282.094 | |
NDJMQXWRZUTNSA-UHFFFAOYSA-N | |
68668352 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F |
C14H17BF2O3 | |
MFCD22419259 | |
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone | |
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures