missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals
$285.89 - $285.89
Chemical Identifiers
| CAS | 10315-03-4 |
|---|---|
| Molecular Formula | C13H17NO·ClH |
| Molecular Weight (g/mol) | 239.74 |
| MDL Number | MFCD00039037 |
| InChI Key | JYDHZOIDIWUHDB-UHFFFAOYSA-N |
| Synonym | 4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride |
| PubChem CID | 2723767 |
| IUPAC Name | 1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride |
| SMILES | CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC134580050
|
Thermo Scientific Chemicals
134580050 |
5 g | Glass bottle |
Each for $285.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10315-03-4 | |
| 239.74 | |
| JYDHZOIDIWUHDB-UHFFFAOYSA-N | |
| 2723767 | |
| CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl |
| C13H17NO·ClH | |
| MFCD00039037 | |
| 4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride | |
| 1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride |
Specifications
| 10315-03-4 | |
| 100.0 | |
| Yellow to Beige | |
| 99% | |
| C13H17NO·ClH | |
| 5 g | |
| JYDHZOIDIWUHDB-UHFFFAOYSA-N | |
| 1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride | |
| 2723767 | |
| 99% | |
| 4-Acetyl-4-phenylpiperidine hydrochloride, 99% |
| 98.5 | |
| 232°C to 234°C | |
| Authentic | |
| Glass bottle | |
| MFCD00039037 | |
| 4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride | |
| CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl | |
| 239.74 | |
| 239.74 | |
| Powder |
Safety and Handling
EINECSNumber : 233-694-0
RUO – Research Use Only