missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetylphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 149104-90-5
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01074667
InChI Key OBQRODBYVNIZJU-UHFFFAOYSA-N
Synonym 4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl, 4acetylphenylboronic acid, 4-actylphenylboronic acid, 4acetylbenzeneboronic acid, p-acetylbenzeneboronic acid
PubChem CID 3702122
IUPAC Name (4-acetylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)C)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369400010
SDS View Documents
Thermo Scientific Chemicals
369400010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC369400050
SDS View Documents
Thermo Scientific Chemicals
369400050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC369400250
SDS View Documents
Thermo Scientific Chemicals
369400250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

149104-90-5
163.97
OBQRODBYVNIZJU-UHFFFAOYSA-N
3702122
B(C1=CC=C(C=C1)C(=O)C)(O)O
C8H9BO3
MFCD01074667
4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl, 4acetylphenylboronic acid, 4-actylphenylboronic acid, 4acetylbenzeneboronic acid, p-acetylbenzeneboronic acid
(4-acetylphenyl)boronic acid

Specifications

149104-90-5
100.0
97%
C8H9BO3
1g
OBQRODBYVNIZJU-UHFFFAOYSA-N
(4-acetylphenyl)boronic acid
3702122
97%
96.0
Authentic
Glass bottle
MFCD01074667
4-acetylphenyl boronic acid, p-acetylphenylboronic acid, 4-acetylbenzeneboronic acid, 1-4-dihydroxyboranyl phenyl ethan-1-one, boronic acid, 4-acetylphenyl, 4acetylphenylboronic acid, 4-actylphenylboronic acid, 4acetylbenzeneboronic acid, p-acetylbenzeneboronic acid
B(C1=CC=C(C=C1)C(=O)C)(O)O
163.97
163.97
4-Acetylphenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only