missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-1-Boc-piperidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3077906

 View more versions of this product

Catalog No. AAH3077906


Explore available promotions

Description

Description

4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

4-Amino-1-Boc-piperidine
44°C to 48°C
MFCD01076201
UN3259
4-amino-1-boc-piperidine, 1-boc-4-aminopiperidine, n-boc-4-aminopiperidine, n-boc-4-amino-piperidine, 4-amino-1-n-boc-piperidine, 4-amino-n-boc piperidine, 1-boc-4-amino piperidine, 1-n-boc-4-aminopiperidine, 1-t-butoxycarbonyl-4-aminopiperidine, n-1-boc-4-aminopiperidine
LZRDHSFPLUWYAX-UHFFFAOYSA-O
tert-butyl 4-aminopiperidine-1-carboxylate
1268291
97%
87120-72-7
C10H21N2O2
5 g
Air Sensitive
Insoluble in water.
CC(C)(C)OC(=O)N1CCC([NH3+])CC1
201.29
200.28

Chemical Identifiers

87120-72-7
201.29
LZRDHSFPLUWYAX-UHFFFAOYSA-O
1268291
CC(C)(C)OC(=O)N1CCC([NH3+])CC1
C10H21N2O2
MFCD01076201
4-amino-1-boc-piperidine, 1-boc-4-aminopiperidine, n-boc-4-aminopiperidine, n-boc-4-amino-piperidine, 4-amino-1-n-boc-piperidine, 4-amino-n-boc piperidine, 1-boc-4-amino piperidine, 1-n-boc-4-aminopiperidine, 1-t-butoxycarbonyl-4-aminopiperidine, n-1-boc-4-aminopiperidine
tert-butyl 4-aminopiperidine-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only