missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,6-dichlorophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1156830

 View more versions of this product

Catalog No. AAA1156830


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Amino-2,6-dichlorophenol
164°C to 169°C
MFCD00007875
2361665
KGEXISHTCZHGFT-UHFFFAOYSA-N
4-amino-2,6-dichlorophenol
80037
97%
5930-28-9
C6H5Cl2NO
250 g
2,6-dichloro-4-aminophenol, phenol, 4-amino-2,6-dichloro, 2,6-dichloro-p-aminophenol, 3,5-dichloro-4-hydroxyaniline, unii-i0l4csx5n0, i0l4csx5n0, 2,6-dichloro-4-amino phenol, 4-amino-2,6-dichloro-phenol, acmc-209mc4, dsstox_cid_20720
NC1=CC(Cl)=C(O)C(Cl)=C1
178.01
178.02

Chemical Identifiers

5930-28-9
178.01
KGEXISHTCZHGFT-UHFFFAOYSA-N
80037
NC1=CC(Cl)=C(O)C(Cl)=C1
C6H5Cl2NO
MFCD00007875
2,6-dichloro-4-aminophenol, phenol, 4-amino-2,6-dichloro, 2,6-dichloro-p-aminophenol, 3,5-dichloro-4-hydroxyaniline, unii-i0l4csx5n0, i0l4csx5n0, 2,6-dichloro-4-amino phenol, 4-amino-2,6-dichloro-phenol, acmc-209mc4, dsstox_cid_20720
4-amino-2,6-dichlorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 227-671-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only