missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

4-Amino-2-methylbenzoic acid, 98%, Thermo Scientific Chemicals

$61.25 - $195.52

Identifiants chimiques

CAS 2486-75-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD06656133
InChI Key XRSQZFJLEPBPOZ-UHFFFAOYSA-N
Synonym 4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
PubChem CID 241632
IUPAC Name 4-amino-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)N)C(=O)O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAH3165703
Afficher les documents
Thermo Scientific Chemicals
H3165703
1 g
chaque for $61.25
 
AAH3165706
Afficher les documents
Thermo Scientific Chemicals
H3165706
5 g
chaque for $195.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

2486-75-1
151.165
XRSQZFJLEPBPOZ-UHFFFAOYSA-N
241632
CC1=C(C=CC(=C1)N)C(=O)O
C8H9NO2
MFCD06656133
4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
4-amino-2-methylbenzoic acid

Spécifications

2486-75-1
MFCD06656133
4-amino-2-methyl-benzoic acid, 2-methyl-4-aminobenzoic acid, 2-methyl-4-amino-benzoic acid, 4-amino-2-methyl benzoic acid, benzoic acid, 4-amino-2-methyl, pubchem10924, 4-amino-o-toluic acid, methyl-p-aminobenzoesaure, acmc-1cnb3, methyl-4-amino benzoic acid
CC1=C(C=CC(=C1)N)C(=O)O
151.165
151.16
Solid
C8H9NO2
1 g
XRSQZFJLEPBPOZ-UHFFFAOYSA-N
4-amino-2-methylbenzoic acid
241632
98%
4-Amino-2-methylbenzoic acid
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only