Learn More
4-Amino-2-methylpyridine, 98%
CAS: 18437-58-6 | C6H8N2 | 108.144 g/mol
$93.04 - $880.89
Chemical Identifiers
| CAS | 18437-58-6 |
|---|---|
| Molecular Formula | C6H8N2 |
| Molecular Weight (g/mol) | 108.144 |
| MDL Number | MFCD00186509 |
| InChI Key | GNCLPNMQEGMNTG-UHFFFAOYSA-N |
| Synonym | 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118 |
| PubChem CID | 728670 |
| IUPAC Name | 2-methylpyridin-4-amine |
| SMILES | CC1=NC=CC(=C1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH55036MD
|
Thermo Scientific Chemicals
H55036MD |
250 mg |
N/A
|
|
|||||
|
AAH5503603
|
Thermo Scientific Chemicals
H5503603 |
1 g |
N/A
|
|
|||||
|
AAH5503606
|
Thermo Scientific Chemicals
H5503606 |
5 g |
N/A
|
|
|||||
Description
4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-2-methylpyridine is used to prepare 2-methyl-pyridin-4-ol; nitrate at 20°C.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 18437-58-6 | |
| 108.144 | |
| GNCLPNMQEGMNTG-UHFFFAOYSA-N | |
| 728670 | |
| CC1=NC=CC(=C1)N |
| C6H8N2 | |
| MFCD00186509 | |
| 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118 | |
| 2-methylpyridin-4-amine |
Specifications
| 18437-58-6 | |
| C6H8N2 | |
| 250 mg | |
| Soluble in methanol. | |
| CC1=NC=CC(=C1)N | |
| 108.144 | |
| 108.14 | |
| 4-Amino-2-methylpyridine |
| 93°C to 97°C | |
| MFCD00186509 | |
| 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118 | |
| GNCLPNMQEGMNTG-UHFFFAOYSA-N | |
| 2-methylpyridin-4-amine | |
| 728670 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only