missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-bromopyridine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 13534-98-0
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD02068297
InChI Key DDQYSZWFFXOXER-UHFFFAOYSA-N
Synonym 4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
PubChem CID 26095
IUPAC Name 3-bromopyridin-4-amine
SMILES C1=CN=CC(=C1N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC360620010
SDS View Documents
Thermo Scientific Chemicals
360620010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC360620050
SDS View Documents
Thermo Scientific Chemicals
360620050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13534-98-0
173.01
DDQYSZWFFXOXER-UHFFFAOYSA-N
26095
C1=CN=CC(=C1N)Br
C5H5BrN2
MFCD02068297
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
3-bromopyridin-4-amine

Specifications

13534-98-0
100.0
97.5% min. (GC)
C5H5BrN2
1g
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
C1=CN=CC(=C1N)Br
173.01
173.01
4-Amino-3-bromopyridine
97.5
Authentic
Glass bottle
MFCD02068297
22, V,9, 185
DDQYSZWFFXOXER-UHFFFAOYSA-N
3-bromopyridin-4-amine
26095
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only