missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-fluorophenol, 96%, Thermo Scientific Chemicals

$141.96 - $337.73

Chemical Identifiers

CAS 399-95-1
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00077452
InChI Key MNPLTKHJEAFOCA-UHFFFAOYSA-N
Synonym 3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79
PubChem CID 2735919
IUPAC Name 4-amino-3-fluorophenol
SMILES C1=CC(=C(C=C1O)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26084MD
View Documents
Thermo Scientific Chemicals
H26084MD
250 mg
Each for $141.96
 
AAH2608403
View Documents
Thermo Scientific Chemicals
H2608403
1 g
Each for $337.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

399-95-1
127.118
MNPLTKHJEAFOCA-UHFFFAOYSA-N
2735919
C1=CC(=C(C=C1O)F)N
C6H6FNO
MFCD00077452
3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79
4-amino-3-fluorophenol

Specifications

399-95-1
C6H6FNO
250 mg
3-fluoro-4-aminophenol, 2-fluoro-4-hydroxyaniline, phenol, 4-amino-3-fluoro, 4-amino-3-fluoro-phenol, pubchem2832, 3-fluoro-4-amino-phenol, 4-amino-3-fluorophenol, acmc-209j9q, 4-amine-3-fluorophenol, buttpark 33\04-79
C1=CC(=C(C=C1O)F)N
127.118
127.12
4-Amino-3-fluorophenol
137°C to 140°C
MFCD00077452
UN3077
MNPLTKHJEAFOCA-UHFFFAOYSA-N
4-amino-3-fluorophenol
2735919
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c

H302-H317-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 402-230-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only