Learn More
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%, Thermo Scientific Chemicals
$81.78 - $236.88
Chemical Identifiers
CAS | 116-63-2 |
---|---|
Molecular Formula | C10H9NO4S |
Molecular Weight (g/mol) | 239.25 |
MDL Number | MFCD00004019 |
InChI Key | RXCMFQDTWCCLBL-UHFFFAOYSA-N |
Synonym | 1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine |
PubChem CID | 8316 |
ChEBI | CHEBI:19024 |
IUPAC Name | 4-amino-3-hydroxynaphthalene-1-sulfonic acid |
SMILES | C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC149270250
|
Thermo Scientific Chemicals
149270250 |
25 g | Glass bottle |
Each for $81.78
|
|
|||||
AC149271000
|
Thermo Scientific Chemicals
149271000 |
100 g | Glass bottle |
Each for $236.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
116-63-2 | |
239.25 | |
RXCMFQDTWCCLBL-UHFFFAOYSA-N | |
8316 | |
4-amino-3-hydroxynaphthalene-1-sulfonic acid |
C10H9NO4S | |
MFCD00004019 | |
1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine | |
CHEBI:19024 | |
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O |
Specifications
116-63-2 | |
100.0 | |
Pink or Purple | |
95% | |
C10H9NO4S | |
MFCD00004019 | |
14, 846 | |
1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine | |
RXCMFQDTWCCLBL-UHFFFAOYSA-N | |
4-amino-3-hydroxynaphthalene-1-sulfonic acid | |
8316 | |
239.25 | |
Granular Powder |
94.0 | |
295.0°C | |
Authentic | |
Glass bottle | |
H2NC10H5(OH)SO3H | |
25 g | |
15, 450 | |
Solubility in water: insoluble. Other solubilities: soluble in hot sodium bisulfite sol., soluble in alkaline solutions, insoluble in alcohol, ether and benzene | |
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O | |
239.25 | |
CHEBI:19024 | |
95% | |
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-147-3
RTECSNumber : QK1292000
TSCA : TSCA
RUO – Research Use Only