Learn More
4-Amino-3-nitrophenol, 98%, Thermo Scientific Chemicals
$60.81 - $186.27
Chemical Identifiers
CAS | 610-81-1 |
---|---|
Molecular Formula | C6H6N2O3 |
Molecular Weight (g/mol) | 154.125 |
MDL Number | MFCD00066310 |
InChI Key | IQXUIDYRTHQTET-UHFFFAOYSA-N |
Synonym | 4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol |
PubChem CID | 3758882 |
IUPAC Name | 4-amino-3-nitrophenol |
SMILES | C1=CC(=C(C=C1O)[N+](=O)[O-])N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0948706
|
Thermo Scientific Chemicals
L0948706 |
5 g |
Each for $60.81
|
|
||||||
AAL0948714
|
Thermo Scientific Chemicals
L0948714 |
25 g |
Each for $186.27
|
|
||||||
Description
4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.
Solubility
Soluble in water. (1.8 g/L.)
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
610-81-1 | |
154.125 | |
IQXUIDYRTHQTET-UHFFFAOYSA-N | |
3758882 | |
C1=CC(=C(C=C1O)[N+](=O)[O-])N |
C6H6N2O3 | |
MFCD00066310 | |
4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol | |
4-amino-3-nitrophenol |
Specifications
610-81-1 | |
Characteristic | |
MFCD00066310 | |
2210196 | |
Soluble in water. (1.8g/L.) | |
C1=CC(=C(C=C1O)[N+](=O)[O-])N | |
154.125 | |
154.13 | |
4-Amino-3-nitrophenol |
148°C to 153°C | |
C6H6N2O3 | |
5 g | |
4-hydroxy-2-nitroaniline, 3-nitro-4-aminophenol, phenol, 4-amino-3-nitro, 4-amino-3-nitro-phenol, unii-r5wy41q95z, 3-nitro-4-amino phenol, 4-aminophenol-3-nitrophenol, pubchem1481, 4-amino-3-nitropbenol | |
IQXUIDYRTHQTET-UHFFFAOYSA-N | |
4-amino-3-nitrophenol | |
3758882 | |
98% |
Safety and Handling
GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H341
EINECSNumber : 210-236-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only