missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
4-Amino-3-(trifluoromethoxy)benzoic acid, 98%, Thermo Scientific Chemicals
$613.23 - $613.23
Identifiants chimiques
CAS | 175278-22-5 |
---|---|
Molecular Formula | C8H6F3NO3 |
Molecular Weight (g/mol) | 221.135 |
MDL Number | MFCD00205143 |
InChI Key | IXJFWBNYFTWBOR-UHFFFAOYSA-N |
Synonym | 4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s |
PubChem CID | 2735951 |
IUPAC Name | 4-amino-3-(trifluoromethoxy)benzoic acid |
SMILES | C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
AAH3291514
|
Thermo Scientific Chemicals
H3291514 |
25 g |
chaque for $613.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
175278-22-5 | |
221.135 | |
IXJFWBNYFTWBOR-UHFFFAOYSA-N | |
2735951 | |
C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N |
C8H6F3NO3 | |
MFCD00205143 | |
4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s | |
4-amino-3-(trifluoromethoxy)benzoic acid |
Spécifications
175278-22-5 | |
MFCD00205143 | |
4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s | |
C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N | |
221.135 | |
221.14 | |
4-Amino-3-(trifluoromethoxy)benzoic acid |
C8H6F3NO3 | |
25 g | |
IXJFWBNYFTWBOR-UHFFFAOYSA-N | |
4-amino-3-(trifluoromethoxy)benzoic acid | |
2735951 | |
98% |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only