missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-3-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3248703

 View more versions of this product

Catalog No. AAH3248703


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Amino-3-(trifluoromethyl)benzoic acid
212°C to 215°C
MFCD03407960
3291493
NPPPORJZPNJXNQ-UHFFFAOYSA-N
4-amino-3-(trifluoromethyl)benzoic acid
3254286
98%
400-76-0
C8H6F3NO2
1 g
4-amino-3-trifluoromethyl benzoic acid, 4-amino-3-trifluoromethylbenzioc acid, rarechem al bo 0877, 4-amino-5-trifluoromethylbenzoic acid, 4-amino-3-trifluoromethyl-benzoic acid, benzoic acid, 4-amino-3-trifluoromethyl, 4-amino-3-trifluoromethyl benzoic acid 3-trifluoromethyl-4-aminobenzoic acid, 4-amino-3-trifluoromethylbenzoicacid, pubchem2211
NC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
205.14
205.14

Chemical Identifiers

400-76-0
205.14
NPPPORJZPNJXNQ-UHFFFAOYSA-N
3254286
NC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
C8H6F3NO2
MFCD03407960
4-amino-3-trifluoromethyl benzoic acid, 4-amino-3-trifluoromethylbenzioc acid, rarechem al bo 0877, 4-amino-5-trifluoromethylbenzoic acid, 4-amino-3-trifluoromethyl-benzoic acid, benzoic acid, 4-amino-3-trifluoromethyl, 4-amino-3-trifluoromethyl benzoic acid 3-trifluoromethyl-4-aminobenzoic acid, 4-amino-3-trifluoromethylbenzoicacid, pubchem2211
4-amino-3-(trifluoromethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only