missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-4'-cyanobiphenyl 95.0+%, TCI America™

$221.46 - $802.76

Chemical Identifiers

CAS 4854-84-6
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00191367
InChI Key CPJQKNUJNWPAPH-UHFFFAOYSA-N
PubChem CID 97193
IUPAC Name 4-(4-aminophenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A12491G
View Documents
TCI America
A12491G
1 g
Each for $221.46
 
A12495G
View Documents
TCI America
A12495G
5 g
Each for $802.76
 
Description

Description

Specifications

Chemical Identifiers

4854-84-6
194.237
CPJQKNUJNWPAPH-UHFFFAOYSA-N
4-(4-aminophenyl)benzonitrile
C13H10N2
MFCD00191367
97193
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)N

Specifications

4854-84-6
Yellow
MFCD00191367
4′C-Aminobiphenyl-4-carbonitrile, 4-Amino-4′C-cyanodiphenyl
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)N
194.237
194.24
Crystalline Powder
185°C
C13H10N2
1 g
CPJQKNUJNWPAPH-UHFFFAOYSA-N
4-(4-aminophenyl)benzonitrile
97193
≥95.0% (GC)
4-Amino-4′-cyanobiphenyl
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DV1763000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'