missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-5-chloro-2,1,3-benzothiadiazole 97.0+%, TCI America™
Supplier: TCI America A15135G
Specifications
4-Amino-5-chloro-2,1,3-benzothiadiazole | |
89°C | |
MFCD00465908 | |
4-Amino-5-chloropiazthiole | |
C1=CC2=NSN=C2C(=C1Cl)N | |
185.629 | |
185.63 | |
Crystalline Powder |
30536-19-7 | |
C6H4ClN3S | |
5 g | |
MURNIACGGUSMAP-UHFFFAOYSA-N | |
5-chloro-2,1,3-benzothiadiazol-4-amine | |
771620 | |
≥97.0% (T) |
Chemical Identifiers
30536-19-7 | |
185.629 | |
MURNIACGGUSMAP-UHFFFAOYSA-N | |
771620 | |
C1=CC2=NSN=C2C(=C1Cl)N |
C6H4ClN3S | |
MFCD00465908 | |
4-Amino-5-chloropiazthiole | |
5-chloro-2,1,3-benzothiadiazol-4-amine |
Safety and Handling
TSCA : No