missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoantipyrine Hydrochloride 98.0+%, TCI America™
$72.70 - $188.27
Chemical Identifiers
| CAS | 22198-72-7 |
|---|---|
| Molecular Formula | C11H14ClN3O |
| Molecular Weight (g/mol) | 239.703 |
| MDL Number | MFCD00042012 |
| InChI Key | UZSCVCWALGRUTR-UHFFFAOYSA-N |
| Synonym | 4-aminoantipyrine hydrochloride, 4-amino-1,5-dimethyl-2-phenyl-1h-pyrazol-3 2h-one hydrochloride, unii-f7wgh82i6d, aminoantipyrine hydrochloride, 4-aminoantipyrine hcl, f7wgh82i6d, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, monohydrochloride, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, hydrochloride 1:1, ampyrone hydrochloride, antipyrine, 4-amino-, monohydrochloride |
| PubChem CID | 72639 |
| IUPAC Name | 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one;hydrochloride |
| SMILES | CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N.Cl |
Chemical Identifiers
| 22198-72-7 | |
| 239.703 | |
| UZSCVCWALGRUTR-UHFFFAOYSA-N | |
| 72639 | |
| CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N.Cl |
| C11H14ClN3O | |
| MFCD00042012 | |
| 4-aminoantipyrine hydrochloride, 4-amino-1,5-dimethyl-2-phenyl-1h-pyrazol-3 2h-one hydrochloride, unii-f7wgh82i6d, aminoantipyrine hydrochloride, 4-aminoantipyrine hcl, f7wgh82i6d, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, monohydrochloride, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, hydrochloride 1:1, ampyrone hydrochloride, antipyrine, 4-amino-, monohydrochloride | |
| 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one;hydrochloride |
Specifications
| 22198-72-7 | |
| MFCD00042012 | |
| 4-aminoantipyrine hydrochloride, 4-amino-1,5-dimethyl-2-phenyl-1h-pyrazol-3 2h-one hydrochloride, unii-f7wgh82i6d, aminoantipyrine hydrochloride, 4-aminoantipyrine hcl, f7wgh82i6d, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, monohydrochloride, 3h-pyrazol-3-one, 4-amino-1,2-dihydro-1,5-dimethyl-2-phenyl-, hydrochloride 1:1, ampyrone hydrochloride, antipyrine, 4-amino-, monohydrochloride | |
| CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N.Cl | |
| 239.703 | |
| 239.70 | |
| Crystalline Powder |
| C11H14ClN3O | |
| 5 g | |
| UZSCVCWALGRUTR-UHFFFAOYSA-N | |
| 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one;hydrochloride | |
| 72639 | |
| ≥98.0% (HPLC,T) | |
| 4-Aminoantipyrine Hydrochloride [for Biochemical Research] |
Safety and Handling
EINECSNumber : (9)-0062
TSCA : Yes