missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Aminobenzohydrazide 98.0+%, TCI America™
Supplier: TCI America A121125G
Specifications
4-Aminobenzohydrazide | |
228°C | |
C7H9N3O | |
25 g | |
WPBZMCGPFHZRHJ-UHFFFAOYSA-N | |
4-aminobenzohydrazide | |
21450 | |
≥98.0% (HPLC,T) |
5351-17-7 | |
White-Yellow | |
MFCD00007606 | |
4-aminobenzhydrazide, amben hydrazide, p-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, aminostimil, 4-aminobenzoylhydrazine, p-aminobenzoic acid hydrazide, p-aminobenzoyl hydrazide, benzoic acid, 4-amino-, hydrazide, p-aminobenzoic hydrazide | |
C1=CC(=CC=C1C(=O)NN)N | |
151.169 | |
151.17 | |
Crystalline Powder |
Chemical Identifiers
5351-17-7 | |
151.169 | |
WPBZMCGPFHZRHJ-UHFFFAOYSA-N | |
21450 | |
C1=CC(=CC=C1C(=O)NN)N |
C7H9N3O | |
MFCD00007606 | |
4-aminobenzhydrazide, amben hydrazide, p-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, aminostimil, 4-aminobenzoylhydrazine, p-aminobenzoic acid hydrazide, p-aminobenzoyl hydrazide, benzoic acid, 4-amino-, hydrazide, p-aminobenzoic hydrazide | |
4-aminobenzohydrazide |
Safety and Handling
EINECSNumber : (3)-0235
RTECSNumber : DG2580000
TSCA : No