missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminobenzoic Acid, 99%, Thermo Scientific Chemicals

$86.27 - $239.63

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146212500
SDS View Documents
Thermo Scientific Chemicals
146212500
250 g Glass bottle
Each for $86.27
 
AC146210010
SDS View Documents
Thermo Scientific Chemicals
146210010
1 kg Plastic Bottle
Each for $239.63
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

150-13-0
137.14
ALYNCZNDIQEVRV-UHFFFAOYSA-N
978
4-aminobenzoic acid
C7H7NO2
MFCD00007894
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
CHEBI:30753
NC1=CC=C(C=C1)C(O)=O

Specifications

150-13-0
100.0
White to Yellow
99%
C7H7NO2
MFCD00007894
14, 418
15, 418
Solubility in water: 4.7g/L (20°C). Other solubilities: soluble in glacial acetic acid,soluble in ethyl acetate,slightly soluble in benzene,121g/L etanol- 1g/50mL ether,soluble in chloroform
NC1=CC=C(C=C1)C(O)=O
137.14
CHEBI:30753
99%
4-Aminobenzoic acid
98.5
186.0°C to 189.0°C
Authentic
Glass bottle
H2NC6H4CO2H
250 g
02,24
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
ALYNCZNDIQEVRV-UHFFFAOYSA-N
4-aminobenzoic acid
978
137.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.

Warning

EINECSNumber : 205-753-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only