missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminohippuric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 153800250

 View more versions of this product

Catalog No. AC153800250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Aminohippuric acid
98.5
198.0°C to 202.0°C
Authentic
Plastic bottle
H2NC6H4CONHCH2CO2H
25 g
15, 439
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, benzene, aceton and most, organic solvents, pract. insoluble in ccl4, very soluble in dilute hydrochloric acid and alkal
C1=CC(=CC=C1C(=O)NCC(=O)O)N
194.19
CHEBI:104011
99%
61-78-9
100.0
Gray to White
99%
C9H10N2O3
MFCD00007890
14, II, 258
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
HSMNQINEKMPTIC-UHFFFAOYSA-N
2-[(4-aminobenzoyl)amino]acetic acid
2148
194.19
Crystalline Powder

Chemical Identifiers

61-78-9
MFCD00007890
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
CHEBI:104011
C1=CC(=CC=C1C(=O)NCC(=O)O)N
194.19
HSMNQINEKMPTIC-UHFFFAOYSA-N
2148
2-[(4-aminobenzoyl)amino]acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-518-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only