missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2351936

 View more versions of this product

Catalog No. AAB2351936

Explore available promotions

Description

Description

4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

Solubility
Slightly soluble in water. Insoluble in ethanol, benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications

Specifications

4-(Aminomethyl)benzoic acid
∼290°C (decomposition)
MFCD00010203
1100606
Slightly soluble in water. Insoluble in ethanol,benzene and chloroform.
C1=CC(=CC=C1CN)C(=O)O
151.165
151.16
56-91-7
C8H9NO2
500 g
4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
QCTBMLYLENLHLA-UHFFFAOYSA-N
4-(aminomethyl)benzoic acid
65526
97%

Chemical Identifiers

56-91-7
151.165
QCTBMLYLENLHLA-UHFFFAOYSA-N
65526
C1=CC(=CC=C1CN)C(=O)O
C8H9NO2
MFCD00010203
4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
4-(aminomethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-297-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only