missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzoic acid, 99%

CAS: 56-91-7 | C8H9NO2 | 151.17 g/mol

$162.21 - $400.27

Chemical Identifiers

CAS 56-91-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00010203
InChI Key QCTBMLYLENLHLA-UHFFFAOYSA-N
Synonym 4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
PubChem CID 65526
IUPAC Name 4-(aminomethyl)benzoic acid
SMILES C1=CC(=CC=C1CN)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC232220250
SDS View Documents
Thermo Scientific Chemicals
232220250
25 g Glass bottle
Each for $162.21
Only null left
 
AC232221000
SDS View Documents
Thermo Scientific Chemicals
232221000
100 g Glass bottle
Each for $400.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-91-7
151.17
QCTBMLYLENLHLA-UHFFFAOYSA-N
65526
C1=CC(=CC=C1CN)C(=O)O
C8H9NO2
MFCD00010203
4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
4-(aminomethyl)benzoic acid

Specifications

56-91-7
100.0
White
Authentic
Glass bottle
H2NCH2C6H4COOH
25 g
QCTBMLYLENLHLA-UHFFFAOYSA-N
4-(aminomethyl)benzoic acid
65526
99%
4-(Aminomethyl)benzoic acid, 99%
98.5
>300.0°C
>300°C
98.5% min. (HPLC)
C8H9NO2
MFCD00010203
4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
C1=CC(=CC=C1CN)C(=O)O
151.17
151.17
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-297-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only