Learn More
4-(Aminomethyl)benzonitrile hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2747306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(Aminomethyl)benzonitrile hydrochloride | |
279°C to 284°C | |
MFCD01861472 | |
4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl | |
C1=CC(=CC=C1CN)C#N.Cl | |
168.624 | |
168.62 |
15996-76-6 | |
C8H9ClN2 | |
5 g | |
QREZLLYPLRPULF-UHFFFAOYSA-N | |
4-(aminomethyl)benzonitrile;hydrochloride | |
9942250 | |
97% |
Chemical Identifiers
15996-76-6 | |
168.624 | |
QREZLLYPLRPULF-UHFFFAOYSA-N | |
9942250 | |
C1=CC(=CC=C1CN)C#N.Cl |
C8H9ClN2 | |
MFCD01861472 | |
4-aminomethyl benzonitrile hydrochloride, 4-cyanobenzylamine hcl, 4-cyanobenzylamine hydrochloride, 4-aminomethyl benzonitrile hcl, 4-aminomethyl-benzonitrile hydrochloride, benzonitrile, 4-aminomethyl-, monohydrochloride, p-aminomethyl benzonitrile hydrochloride, pubchem2057, acmc-209dkg, 4-aminomethylbenzonitrile hcl | |
4-(aminomethyl)benzonitrile;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only