missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Aminooctanophenone 97.0+%, TCI America™

Supplier:  TCI America A184925G

Catalog No. A184925G


Explore available promotions

Description

Description

Specifications

Specifications

4′-Aminooctanophenone
105°C
C14H21NO
25 g
IMQPGJKTCYTPEI-UHFFFAOYSA-N
1-(4-aminophenyl)octan-1-one
44965
≥97.0% (GC,T)
63884-78-6
White-Yellow
MFCD01737717
4-n-Octanoylaniline
CCCCCCCC(=O)C1=CC=C(N)C=C1
219.33
219.33
Crystalline Powder

Chemical Identifiers

63884-78-6
219.33
IMQPGJKTCYTPEI-UHFFFAOYSA-N
44965
CCCCCCCC(=O)C1=CC=C(N)C=C1
C14H21NO
MFCD01737717
4-n-Octanoylaniline
1-(4-aminophenyl)octan-1-one
Videos
Safety and Handling

Safety and Handling

RTECSNumber : EY4375000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions