missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminophenol 98.0+%, TCI America™

Supplier:  TCI America A038425G

 View more versions of this product

Catalog No. A038425G


Explore available promotions

Description

Description

Specifications

Specifications

4-Aminophenol
187°C
C6H7NO
25 g
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
C1=CC(=CC=C1N)O
109.128
CHEBI:17602
≥98.0% (GC,T)
123-30-8
284°C
MFCD00007869
2512
PLIKAWJENQZMHA-UHFFFAOYSA-N
4-aminophenol
403
109.13
Crystalline Powder

Chemical Identifiers

123-30-8
109.128
PLIKAWJENQZMHA-UHFFFAOYSA-N
403
4-aminophenol
C6H7NO
MFCD00007869
p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
CHEBI:17602
C1=CC(=CC=C1N)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0675

RTECSNumber : SJ5075000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions