missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminoquinoline, 95+%, Thermo Scientific™

Chemical Identifiers

CAS 578-68-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.17
InChI Key FQYRLEXKXQRZDH-UHFFFAOYSA-N
Synonym 4-aminoquinoline, 4-quinolinamine, 4-aminoguinoline, quinolin-4-ylamine, 4-quinolylamine, unii-gte5p5l97n, ccris 1679, gte5p5l97n, 4-quinolinylamine, 4-amino quinoline
PubChem CID 68476
IUPAC Name quinolin-4-amine
SMILES C1=CC=C2C(=C1)C(=CC=N2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC459762500
View Documents
Thermo Scientific Chemicals
459762500
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC459760010
View Documents
Thermo Scientific Chemicals
459760010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

578-68-7
144.17
4-aminoquinoline, 4-quinolinamine, 4-aminoguinoline, quinolin-4-ylamine, 4-quinolylamine, unii-gte5p5l97n, ccris 1679, gte5p5l97n, 4-quinolinylamine, 4-amino quinoline
quinolin-4-amine
C9H8N2
FQYRLEXKXQRZDH-UHFFFAOYSA-N
68476
C1=CC=C2C(=C1)C(=CC=N2)N

Specifications

578-68-7
Authentic
C9H8N2
4-aminoquinoline, 4-quinolinamine, 4-aminoguinoline, quinolin-4-ylamine, 4-quinolylamine, unii-gte5p5l97n, ccris 1679, gte5p5l97n, 4-quinolinylamine, 4-amino quinoline
FQYRLEXKXQRZDH-UHFFFAOYSA-N
quinolin-4-amine
68476
95+%
332.0°C to 337.0°C
95% min. (HPLC)
250mg
Solubility in water: slightly soluble.
C1=CC=C2C(=C1)C(=CC=N2)N
144.17
144.17
4-Aminoquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only