missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminosalicylic acid, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 104625000

 View more versions of this product

Catalog No. AC104625000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Aminosalicylic acid
65-49-6
100.0
99%
C7H7NO3
MFCD00007789
14, 579
4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4-amino-2-hydroxybenzoic acid
4649
153.14
99%
98.5
Authentic
Glass bottle
H2NC6H3(OH)CO2H
500g
15, 473
Solubility in water: 2g/L (20°C). Other solubilities: soluble in dilute hno3 and dilute naoh,aceton,1g/21mL alcohol,slightly soluble in ether,practically insoluble in benzene,chloroform,,carbontetrachloride
C1=CC(=C(C=C1N)O)C(=O)O
153.14
CHEBI:27565
99%

Chemical Identifiers

65-49-6
153.14
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4649
4-amino-2-hydroxybenzoic acid
C7H7NO3
MFCD00007789
4-aminosalicylic acid, aminosalicylic acid, p-aminosalicylic acid, rezipas, pamisyl, parasalindon, aminopar, deapasil, ferrosan, apacil
CHEBI:27565
C1=CC(=C(C=C1N)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-613-5

RTECSNumber : VO1225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only