missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminostilbene 97.0+%, TCI America™

$649.78 - $3134.32

Chemical Identifiers

CAS 834-24-2
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00025377
InChI Key VFPLSXYJYAKZCT-VOTSOKGWSA-N
Synonym 4-Stilbenamine
PubChem CID 1272788
IUPAC Name 4-[(1E)-2-phenylethenyl]aniline
SMILES NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A08545G
View Documents
TCI America
A08545G
5 g
Each for $649.78
 
A085425G
View Documents
TCI America
A085425G
25 g
Each for $3,134.32
 
Description

Description

Specifications

Chemical Identifiers

834-24-2
195.27
VFPLSXYJYAKZCT-VOTSOKGWSA-N
1272788
NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
C14H13N
MFCD00025377
4-Stilbenamine
4-[(1E)-2-phenylethenyl]aniline

Specifications

834-24-2
Yellow
MFCD00025377
4-Stilbenamine
NC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
195.27
195.27
Crystalline Powder
151°C
C14H13N
5 g
VFPLSXYJYAKZCT-VOTSOKGWSA-N
4-[(1E)-2-phenylethenyl]aniline
1272788
≥97.0% (HPLC,T)
4-Aminostilbene
Videos
Safety and Handling

Safety and Handling

RTECSNumber : WJ3500000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'