missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Anilino-1,1':4',1″-terphenyl 98.0+%, TCI America™
Supplier: TCI America A29031G
Specifications
4-Anilino-1,1′:4′,1′′-terphenyl | |
196°C | |
1 g | |
VWXSLLOSYCKNCF-UHFFFAOYSA-N | |
N-phenyl-4-(4-phenylphenyl)aniline | |
17826786 | |
≥98.0% (HPLC,N) |
897671-81-7 | |
C24H19N | |
4-Anilino-p-terphenyl | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)NC4=CC=CC=C4 | |
321.423 | |
321.42 | |
Crystalline Powder |
Chemical Identifiers
897671-81-7 | |
321.423 | |
4-Anilino-p-terphenyl | |
N-phenyl-4-(4-phenylphenyl)aniline |
C24H19N | |
VWXSLLOSYCKNCF-UHFFFAOYSA-N | |
17826786 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)NC4=CC=CC=C4 |
Safety and Handling
TSCA : No