missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Benzoyl-4'-bromobiphenyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1848214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Benzoyl-4'-bromobiphenyl | |
155°C to 158°C | |
MFCD00209630 | |
2378429 | |
ITMLYLPKSFZCJK-UHFFFAOYSA-N | |
[4-(4-bromophenyl)phenyl]-phenylmethanone | |
478901 | |
99% |
63242-14-8 | |
C19H13BrO | |
25 g | |
4-benzoyl-4'-bromobiphenyl, 4-4-bromophenyl benzophenone, 4'-bromobiphenyl-4-yl phenyl methanone, 4'-bromo-1,1'-biphenyl-4-yl phenyl methanone, 4-4-bromophenyl phenyl-phenyl-methanone, 4-4-bromophenyl phenyl phenyl ketone, 4-4-bromophenyl phenyl-phenylmethanone, 4-4-bromophenyl phenyl phenyl methanone, pubchem9089 | |
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C3=CC=C(C=C3)Br | |
337.216 | |
337.22 |
Chemical Identifiers
63242-14-8 | |
337.216 | |
ITMLYLPKSFZCJK-UHFFFAOYSA-N | |
478901 | |
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C3=CC=C(C=C3)Br |
C19H13BrO | |
MFCD00209630 | |
4-benzoyl-4'-bromobiphenyl, 4-4-bromophenyl benzophenone, 4'-bromobiphenyl-4-yl phenyl methanone, 4'-bromo-1,1'-biphenyl-4-yl phenyl methanone, 4-4-bromophenyl phenyl-phenyl-methanone, 4-4-bromophenyl phenyl phenyl ketone, 4-4-bromophenyl phenyl-phenylmethanone, 4-4-bromophenyl phenyl phenyl methanone, pubchem9089 | |
[4-(4-bromophenyl)phenyl]-phenylmethanone |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only