missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Benzylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5248603

Catalog No. AAH5248603


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-(Benzylcarbamoyl)benzeneboronic acid
212°C to 218°C
MFCD03411953
4-n-benzylaminocarbonyl phenylboronic acid, 4-benzylcarbamoyl phenyl boronic acid, 4-benzylcarbamoyl phenylboronic acid, 4-benzylcarbamoyl benzeneboronic acid, 4-benzylamino-1-carbonyl phenyl boronic acid, 4-n-benzylaminocarbonyl benzeneboronic acid, 4-benzylaminocarbonyl phenylboronic acid, acmc-1bmt1, 4-n-acetyl aminopiperidine
B(C1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2)(O)O
255.08
255.08
252663-47-1
C14H14BNO3
1 g
AAUCAVUVHBOOPF-UHFFFAOYSA-N
[4-(benzylcarbamoyl)phenyl]boronic acid
2773243
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions