missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Benzyloxy-2(1H)-pyridone, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 53937-02-3 |
---|---|
Molecular Formula | C12H11NO2 |
Molecular Weight (g/mol) | 201.23 |
MDL Number | MFCD04039771 |
InChI Key | DOVNUEPFPBWTSV-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one |
PubChem CID | 3607724 |
IUPAC Name | 4-(benzyloxy)-1,2-dihydropyridin-2-one |
SMILES | O=C1NC=CC(OCC2=CC=CC=C2)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC438090050
|
Thermo Scientific Chemicals
438090050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
53937-02-3 | |
201.23 | |
DOVNUEPFPBWTSV-UHFFFAOYSA-N | |
3607724 | |
O=C1NC=CC(OCC2=CC=CC=C2)=C1 |
C12H11NO2 | |
MFCD04039771 | |
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one | |
4-(benzyloxy)-1,2-dihydropyridin-2-one |
Specifications
53937-02-3 | |
100.0 | |
97.5% min. (HPLC) | |
MFCD04039771 | |
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one | |
O=C1NC=CC(OCC2=CC=CC=C2)=C1 | |
201.23 | |
201.23 | |
4-Benzyloxy-2(1H)-pyridone |
97.5 | |
Authentic | |
C12H11NO2 | |
5g | |
DOVNUEPFPBWTSV-UHFFFAOYSA-N | |
4-(benzyloxy)-1,2-dihydropyridin-2-one | |
3607724 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO â Research Use Only