missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Benzyloxy)benzyl Chloride 98.0+%, TCI America™
$184.54 - $548.03
Chemical Identifiers
| CAS | 836-42-0 |
|---|---|
| Molecular Formula | C14H13ClO |
| Molecular Weight (g/mol) | 232.707 |
| MDL Number | MFCD00000916 |
| InChI Key | UYQPSKUPEXAQRJ-UHFFFAOYSA-N |
| Synonym | Benzyl 4-(Chloromethyl)phenyl Ether, 1-(Benzyloxy)-4-(chloromethyl)benzene |
| PubChem CID | 70042 |
| IUPAC Name | 1-(chloromethyl)-4-phenylmethoxybenzene |
| SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)CCl |
Chemical Identifiers
| 836-42-0 | |
| 232.707 | |
| UYQPSKUPEXAQRJ-UHFFFAOYSA-N | |
| 70042 | |
| C1=CC=C(C=C1)COC2=CC=C(C=C2)CCl |
| C14H13ClO | |
| MFCD00000916 | |
| Benzyl 4-(Chloromethyl)phenyl Ether, 1-(Benzyloxy)-4-(chloromethyl)benzene | |
| 1-(chloromethyl)-4-phenylmethoxybenzene |
Specifications
| 836-42-0 | |
| C14H13ClO | |
| 1 g | |
| Benzyl 4-(Chloromethyl)phenyl Ether, 1-(Benzyloxy)-4-(chloromethyl)benzene | |
| C1=CC=C(C=C1)COC2=CC=C(C=C2)CCl | |
| 232.707 | |
| 232.71 | |
| Crystalline Powder |
| 79°C | |
| MFCD00000916 | |
| 3261 | |
| UYQPSKUPEXAQRJ-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-phenylmethoxybenzene | |
| 70042 | |
| ≥98.0% (GC) | |
| 4-(Benzyloxy)benzyl Chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator