missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals

$411.62 - $411.62

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169782500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
169782500
250 g Glass bottle
Each for $411.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-16-2
200.24
VYQNWZOUAUKGHI-UHFFFAOYSA-N
7638
4-phenylmethoxyphenol
C13H12O2
MFCD00002333
4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
CHEBI:34380
C1=CC=C(C=C1)COC2=CC=C(C=C2)O

Specifications

103-16-2
100.0
Brown to White
97.5% min. (GC)
C13H12O2
MFCD00002333
06,845
4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
VYQNWZOUAUKGHI-UHFFFAOYSA-N
4-phenylmethoxyphenol
7638
200.24
Powder, Crystals or Chunks
97.5
121.0°C to 122.0°C
Authentic
Glass bottle
C6H5CH2OC6H4OH
250 g
15,6334
Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,benzene,diethyl ether,,soluble in acetone and alkalies,,practically insoluble in petroleum hydrocarbons
C1=CC=C(C=C1)COC2=CC=C(C=C2)O
200.24
CHEBI:34380
98%
4-(Benzyloxy)phenol, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-083-3

RTECSNumber : SJ7700000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only